Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.31620

Sigma-Aldrich

APT1 Inhibitor II, Cpd21

Synonym(s):

APT1 Inhibitor II, Cpd21, LYPLA1 Inhibitor, Lysophospholipase Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$217.00

$217.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$217.00

About This Item

Empirical Formula (Hill Notation):
C18H17ClF3N3O3
CAS Number:
Molecular Weight:
415.79
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$217.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

840 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(c(cc1)Cl)NC(=O)CN2CCN(CC2)C(=O)c3[o]ccc3

InChI

1S/C18H17ClF3N3O3/c19-13-4-3-12(18(20,21)22)10-14(13)23-16(26)11-24-5-7-25(8-6-24)17(27)15-2-1-9-28-15/h1-4,9-10H,5-8,11H2,(H,23,26)

InChI key

OXKNHBBDOIMFFQ-UHFFFAOYSA-N

General description

A cell permeable acetylpiperazine derivative compound that acts as a selective and reversible inhibitor of lysophospholipase 1 (LYPLA1; IC50 = 840 nM, Ki = 300 nM) without affecting the activity of LYPLA2 and 25 other serine hydrolases in mouse BW5147 T cell hydridoma proteome. Shown to cause an almost complete inhibition (+90%) of LYPLA1 activity in lung, heart, and kidney (50 mg/kg, 3 h), and in HEK293T, and mouse T cells (~ 5 µM for 3 h).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
APT1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Adibekian, A., et al. 2012. J. Am. Chem. Soc.134, 10345.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service