Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.31005

Sigma-Aldrich

PFKFB3 Inhibitor II, PFK15

Synonym(s):

PFKFB3 Inhibitor II, PFK15, ( E)-1-(Pyridin-4-yl)-3-(quinolin-2-yl)prop-2-en-1-one, 1-(Pyridin-4-yl)-3-(quinolin-2-yl)prop-2-en-1-one, 6-Phosphofructo-2-kinase/fructose-2,6-bisphosphatase 3 Inhibitor, PFK15, (E)-1-(Pyridin-4-yl)-3-(quinolin-2-yl)prop-2-en-1-one, 1-(Pyridin-4-yl)-3-(quinolin-2-yl)prop-2-en-1-one, 6-Phosphofructo-2-kinase/fructose-2,6-bisphosphatase 3 Inhibitor, PFK15

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12N2O
CAS Number:
Molecular Weight:
260.29
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, bioavailable and more potent quinolinyl analog of PFKFB3 Inhibitor, 3PO (Cat. No. 525330) that inhibits the activity of 6-Phosphofructo-2-kinase (PFKFB3) by reversibly docking to its substrate binding domain (IC50 = 207 nM compared to 22.9 µM for 3PO). Shown to effectively reduce fructose 2,6-bisphosphate, glucose uptake, and intracellular ATP levels. Does neither affect the activity of 96 other kinases in any significant manner (at ~10 µM) nor PFK-1, hexokinase, phosphoglucoisomerase, or PFKFB4 even at 100 µM. Induces apoptosis and reduces proliferation in transformed cells (mean GI50 of ~3 µM in the NCI-60 cancer cell panel). Displays adequate pharmacokinetic properties to suppress glucose uptake and the growth of Lewis lung carcinoma is syngenic mice and block the growth of human tumor xenografts in athymic mice (~25 mg/kg, i.p. administered every 3rd day).
A cell-permeable, bioavailable and more potent quinolinyl analog of PFKFB3 Inhibitor, 3PO (Cat. No. 525330) that inhibits the activity of 6-Phosphofructo-2-kinase (PFKFB3) by reversibly docking to its substrate binding domain (IC50 = 207 nM compared to 22.9 µM for 3PO). Shown to effectively reduce fructose 2,6-bisphosphate, glucose uptake, and intracellular ATP levels. Does neither affect the activity of 96 other kinases in any significant manner (at ~10 µM) nor PFK-1, hexokinase, phosphoglucoisomerase, or PFKFB4 even at 100 µM. Induces apoptosis and reduces proliferation in transformed cells (mean GI50 of ~3 µM in the NCI-60 cancer cell panel). Displays adequate pharmacokinetic properties to suppress glucose uptake and the growth of Lewis lung carcinoma is syngenic mice and block the growth of human tumor xenografts in athymic mice (~25 mg/kg, i.p. administered every 3rd day).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PFKFB3
Reversible: yes
Target IC50: 207 nM for hrPFKFP3

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 6 months at -70°C.
Use only fresh DMSO for reconstitution.

Other Notes

Clem, B. F., et al. 2013. Mol. Cancer Ther.12, 1461.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service