Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30923

Sigma-Aldrich

Kir6.2/SUR1 Channel Activator, VU0071063

Synonym(s):

Kir6.2/SUR1 Channel Activator, VU0071063, 1,3-Dimethyl-7-(4-(2-methyl-2-propanyl)benzyl)-3,7-dihydro-1H-purine-2,6-dione, 7-(4-( tert-Butyl)benzyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione, Kir6.2/SUR1 Channel Opener, SUR1/Kir6.2 selective K-ATP Channel Opener, 1,3-Dimethyl-7-(4-(2-methyl-2-propanyl)benzyl)-3,7-dihydro-1H-purine-2,6-dione, 7-(4-(tert-Butyl)benzyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione, Kir6.2/SUR1 Channel Opener, SUR1/Kir6.2 selective K-ATP Channel Opener

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22N4O2
CAS Number:
Molecular Weight:
326.39
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A xanthine derivative that serves as a fast acting, reversible, and selective activator of Kir6.2/SUR1 channels (EC50 = 7 µM). Directly occupies the binding site located within SUR1. Shown to be a more potent activator of Kir6.2/SUR1 than diazoxide (EC50 = 120 µM). However, it does not affect Kir6.1/SURA2A or Kir6.2/SURA2A channels and is inactive against Kir2.1 and Kir2.2 (IC50 >100 µM) and exhibits very weak inhibitory activity against Kir3.1/3.2 (IC50 = 65 µM) and Kir2.3 (IC50 = 91 µM) channels. Inhibits glucose-stimulated Ca2+ influx into mouse pancreatic β-cells.
A xanthine derivative that serves as a fast acting, reversible, and selective activator of Kir6.2/SUR1 channels (EC50 = 7 µM). Directly occupies the binding site located within SUR1. Shown to be a more potent activator of Kir6.2/SUR1 than diazoxide (EC50 = 120 µM). However, it does not affect Kir6.1/SURA2A or Kir6.2/SURA2A channels and is inactive against Kir2.1 and Kir2.2 (IC50 >100 µM) and exhibits very weak inhibitory activity against Kir3.1/3.2 (IC50 = 65 µM) and Kir2.3 (IC50 = 91 µM) channels. Inhibits glucose-stimulated Ca2+ influx into mouse pancreatic β-cells.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

EC50 = 7 µM for Kir6.2/SUR1 channel activation
Primary Target
SUR1-containing channels
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Raphemot, R., et al. 2014. Mol. Pharmacol.85, 858.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service