Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30838

Sigma-Aldrich

NMD Inhibitor

≥95% (HPLC), powder, NMD inhibitor, Calbiochem®

Synonym(s):

NMD Inhibitor, Ethyl 2-(((6,7-dimethyl-3-oxo-1,2,3,4-tetrahydro-2-quinoxalinyl)acetyl)amino)-4,5-dimethyl-3-thiophenecarboxylate, Nonsense-Mediated mRNA Decay Inhibitor 14, NMDI-14

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$234.00

$234.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$234.00

About This Item

Empirical Formula (Hill Notation):
C21H25N3O4S
CAS Number:
Molecular Weight:
415.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$234.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

NMD Inhibitor,

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

storage temp.

−20°C

SMILES string

[s]1c(c(c(c1C)C)C(=O)OCC)NC(=O)CC2Nc3c(cc(c(c3)C)C)NC2=O

InChI

1S/C21H25N3O4S/c1-6-28-21(27)18-12(4)13(5)29-20(18)24-17(25)9-16-19(26)23-15-8-11(3)10(2)7-14(15)22-16/h7-8,16,22H,6,9H2,1-5H3,(H,23,26)(H,24,25)

InChI key

QRLPSNLBUMVXBL-UHFFFAOYSA-N

General description

A cell-permeable tetrahydroquinoxalinyl-thiophenecarboxylate compound that inhibits nonsense-mediated mRNA decay (NMD) in a dose-dependent manner and enhances the stability of premature termination codon (PTC) mutated p53 mRNA in N417 and HDQP-1 cells. However, it does not affect the stability of wild-type p53 in U2OS cells. Shown to dock reversibly within a SMG7 pocket and disrupt SMG7-UPF1 interaction and prevent their complex formation. Acts synergistically with G418 (Cat. No. 345810; 200 µg/ml) to increase p53 expression in p53 PTC mutant cells leading to increase mRNA levels of p21, Bax, and PUMA. Does not display any toxicity (>50 µM) in multiple cell lines and has no effect on proliferation and protein synthesis in transformed and non-transformed cells.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
SMG7
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Martin, L., et al. 2014. Cancer Res.74, 3104.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service