Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30750

Sigma-Aldrich

STAT3 Inhibitor XX, APT STAT3-9R

Synonym(s):

STAT3 Inhibitor XX, APT STAT3-9R, H₂N- HGFQWPG(SWTWENGKWTWK)G AYQFLK-GGGGS-RRRRRRRRR-CO₂H

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$291.00

$291.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$291.00

About This Item

Empirical Formula (Hill Notation):
C223H330N80O51
Molecular Weight:
4947.51
UNSPSC Code:
12352200
NACRES:
NA.77

$291.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

H2O: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable STAT3 inhibiting peptide that corresponds to the high affinity STAT3-interacting aptide APTSTAT3 (KD = 231 nM), originally identified by phage peptide library screening, with the addition of 9 arginines at the C-terminus via a GGGGS linker sequence. Reported to inhibit JAK2-catalyzed STAT3 phosphorylation in cell-free kinase assays and suppress cellular STAT3 phosphorylation as well as DNA-binding activity (by 54% post 6 h 30 µM treatment) in human lung carcinoma A549 cultures, resulting in downregulation of cell cycle progression regulator cyclin D1 and reduced expression of anti-apoptic survivin & Bcl-xL, while exhibiting little effect toward cellular phosphorylation levels of STAT1/5 and Akt. Shown to inhibit A549 proliferation (GI50 = 10 to 20 µM in 12 h by MTT assay; complete inhibition by 30 µM in 2-wk colony formation assay) via apoptosis induction (PI+/Annexin V+ population = 15% vs. 2.8% with or without 6 h 30 µM drug treatment) in vitro and intratumoral injection is efficacious in suppressing the growth of established tumors derived from B16F1 murine melanoma allograft as well as from human U87MG glioblastoma and A549 (8 mg/kg, q.o.d.) xenografts in mice in vivo. Binding study indicates that the majority of APTSTAT3 affinity is mediated via STAT2 SH2 domain interaction and only moderate affinity is detected toward DNA-binding domain (DBD).
A cell-permeable, 26-mer peptide (APTSTAT3) that binds to STAT3 with high affinity (Kd = 231 nM) and blocks its DNA binding activity.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
STAT3

Warning

Toxicity: Standard Handling (A)

Sequence

H-His-Gly-Phe-Gln-Trp-Pro-Gly-(Ser-Trp-Thr-Trp-Glu-Asn-Gly-Lys-Trp-Thr-Trp-Lys)-Gly-Ala-Tyr-Gln-Phe-Leu-Lys-Gly-Gly-Gly-Gly-Ser-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kim, D., et al. 2014. Cancer Res.74, 2144.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daejin Kim et al.
Cancer research, 74(8), 2144-2151 (2014-03-01)
STAT3 promotes the survival, proliferation, metastasis, immune escape, and drug resistance of cancer cells, making its targeting an appealing prospect. However, although multiple inhibitors of STAT3 and its regulatory or effector pathway elements have been developed, bioactive agents have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service