Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30746

Sigma-Aldrich

Peroxiredoxin Inhibitor, Conoidin A

Synonym(s):

Peroxiredoxin Inhibitor, Conoidin A, 2,3- bis(Bromomethyl)quinoxaline-1,4-dioxide, Prx Inhibitor, 2,3-bis(Bromomethyl)quinoxaline-1,4-dioxide, Prx Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$290.00

$290.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$290.00

About This Item

Empirical Formula (Hill Notation):
C10H8Br2N2O2
CAS Number:
Molecular Weight:
347.99
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$290.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95% (NMR)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

yellow-brown

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

SMILES string

BrCC1=C(N(c2c(cccc2)[N+]1=O)[O-])CBr

InChI

1S/C10H8Br2N2O2/c11-5-9-10(6-12)14(16)8-4-2-1-3-7(8)13(9)15/h1-4H,5-6H2

InChI key

DQKNFTLRMZOAMG-UHFFFAOYSA-N

General description

A cell-permeable dibromomethylquinoxaline-dioxide compound with nematicidal properties that covalently binds to the peroxidatic cysteine of peroxiredoxin II in Toxoplasma gondii (TgPrxII) and irreversibly blocks its hyperperoxidation activity (IC50 = 23 µM). Also shown to alkylate or crosslink catalytic cysteines of wild type AcePrx-1 in Ancylostoma ceylanicum and human PrxII and PrxIV to inhibit their catalytic activity with similar efficiency. Ineffective towards mitochondrial hPrxIII.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Peroxiredoxin
Reversible: no
Target IC50: 23 µM for blocking TgPrxII hyperperoxidation activity

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 2 weeks at -70°C.
Use only fresh DMSO for reconstitution.

Other Notes

Nguyen, J.B., et al. 2013. Chem. Biol.20, 991.
Haraldsen, J. D., et al. 2009. Org. Biomol. Chem.7, 3040.
Carey, K.L., et al. 2004. Proc. Natl. Acad. Sci. USA101, 7433.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service