Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30616

Sigma-Aldrich

ACE2 Inhibitor, MLN-4760

≥97% (HPLC), powder, Calbiochem®

Synonym(s):

ACE2 Inhibitor, MLN-4760, ( S, S)-2-(1-Carboxy-2-(3-(3,5-dichlorobenzyl)-3H-imidazol-4-yl)-ethylamino)-4-methylpentanoic acid, 2( S)-(1( S)-Carboxy-2-(3-(3,5-dichlorobenzyl)-3H-imidazol-4-yl)-ethylamino)-4-methylpentanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$333.00

$333.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 MG
$333.00

About This Item

Empirical Formula (Hill Notation):
C19H23Cl2N3O4
CAS Number:
Molecular Weight:
428.31
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$333.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

technique(s)

inhibition assay: suitable

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

Clc1cc(cc(c1)C[n]2cncc2C[C@H](N[C@@H](CC(C)C)C(=O)O)C(=O)O)Cl

InChI

1S/C19H23Cl2N3O4/c1-11(2)3-16(18(25)26)23-17(19(27)28)7-15-8-22-10-24(15)9-12-4-13(20)6-14(21)5-12/h4-6,8,10-11,16-17,23H,3,7,9H2,1-2H3,(H,25,26)(H,27,28)/t16-,17-/m0/s1

InChI key

NTCCRGGIJNDEAB-IRXDYDNUSA-N

General description

MLN-4760 is a highly potent and selective cell-permeable inhibitor of angiotensin converting enzyme 2, which is its primary target. (ACE2; IC50 = 440 pM against soluble human ACE2).

Biochem/physiol Actions

MLN-4760 is bioavailable and exhibits far greater selectivity for carboxypeptidase (IC50 = 0.44 nM against 50 pM human ACE2; [ZnCl2] = 10 μM, [MCA-APK(DNP)] = 50 μM) over bovine carboxypeptidase A or ACE peptidyldipeptidase activity (IC50 = 27 μM and >100 μM against 0.5 nM bovine CPDA and 1 nM porcine ACE, respectively; [Substrate] = 50 μM) and porcine ACE (IC50 = 27 and >100 μM, respectively).[1] This reversible ACE2 inhibitor binds to the active site zinc with high-affinity and emulates the transition state during peptide hydrolysis.[2] It reduces serum and kidney ACE 2 activity and abolishes angiotensin II-induced hypertension in mice. MLN-4760 selectively blocks angiotensin (ANG-(1-7)) formation in ACE2 wild type (WT) mice subjected to low ANG II concentrations (<0.1 μM), but at higher ANG II concentrations it does not affect ANG -(1-7) levels in mice. This ACE2 inhibitor enhances tumor necrosis factor (TNF) (10 pg/mL) stimulated expression of proinflammatory cytokines in murine endothelial cells, (1 μM using SVEC-40 line and primary aorta endothelial cultures) in vitro. MLN-4760 is widely employed for studying ACE2 involvement in kidney, cardiovascular and inflammatory bowel diseases via drinking water (10 mg/kg/d), i.v. (0.1 mg/kg), and s.c. (30 mg/kg/d to 300 mg/kg/12 h) injection in rats and mice, in vivo. The inhibitor leucine moiety is shown to simultaneously target ACE2 substrate S1 pocket with its isobutyl group and active site zinc via its carboxylate, while the compound′s 3,5-dichlorobenzyl group effectively occupy S1′ subsite.
MLN-4760 also enables to study the effect of reduced ACE2 activity on the lung′s susceptibility for Coronavirus disease (COVID) related acute respiratory distress syndrome (ARDS).[2]

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John J Byrnes et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 58(11), 819-827 (2009-06-12)
Angiotensin-converting enzyme 2 (ACE2) is expressed in gastrointestinal tissue. Previous studies of GL1001, a potent and selective ACE2 inhibitor, have revealed anti-inflammatory activity in the mouse digestive tract. We hypothesized that GL1001 might also produce beneficial effects in a mouse
Aaron J Trask et al.
American journal of hypertension, 23(6), 687-693 (2010-03-20)
Emerging evidence suggests that cardiac angiotensin-converting enzyme 2 (ACE2) may contribute to the regulation of heart function and hypertension-induced cardiac remodeling. We tested the hypothesis that inhibition of ACE2 in the hearts of (mRen2)27 hypertensive rats may accelerate progression of
Minghao Ye et al.
Hypertension (Dallas, Tex. : 1979), 60(3), 730-740 (2012-07-11)
A newly produced murine recombinant angiotensin (Ang)-converting enzyme 2 (ACE2) was characterized in vivo and in vitro. The effects of available ACE2 inhibitors (MLN-4760 and 2 conformational variants of DX600, linear and cyclic) were also examined. When murine ACE2 was
Paul Towler et al.
The Journal of biological chemistry, 279(17), 17996-18007 (2004-02-03)
The angiotensin-converting enzyme (ACE)-related carboxypeptidase, ACE2, is a type I integral membrane protein of 805 amino acids that contains one HEXXH + E zinc-binding consensus sequence. ACE2 has been implicated in the regulation of heart function and also as a
M J Soler et al.
Kidney international, 72(5), 614-623 (2007-06-21)
Angiotensin converting enzyme 2 (ACE2) is localized to the glomerular epithelial cells. Since ACE2 promotes the degradation of angiotensin II, a decrease in ACE2 activity could lead to the development of glomerular injury. We gave a specific ACE2 inhibitor, MLN-4760

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service