Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.30615

Sigma-Aldrich

GFRα/RET Signaling Activator, XIB-4035

Synonym(s):

GFRα/RET Signaling Activator, XIB-4035, ( E)-N⁴-(7-Chloro-2-(2-chlorostyryl)quinolin-4-yl)-N¹,N¹-diethylpentane-1,4-diamine, N-(7-Chloro-2-(2-(2-chlorophenyl)vinyl)-4-quinolinyl)-N-(4-(diethylamino)-1-methylbutyl)amine, XIB 4035

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H31Cl2N3
CAS Number:
Molecular Weight:
456.45
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

InChI

1S/C26H31Cl2N3/c1-4-31(5-2)16-8-9-19(3)29-26-18-22(14-12-20-10-6-7-11-24(20)28)30-25-17-21(27)13-15-23(25)26/h6-7,10-15,17-19H,4-5,8-9,16H2,1-3H3,(H,29,30)/b14-12+

InChI key

CMLVHSVFSYNMGM-WYMLVPIESA-N

General description

A cell-permeable chlorostyrylquinolinylbutylamino compound that was shown to competitively bind to glial cell line-derived neurotrophic factor (GDNF) family receptor-α1 (GFRα1) and act as an agonist (IC50 = 10.4 µM for 125I-GDNF binding in Neuro-2A cells) and induce autophosphorylation of RET protein. Elicits neurite outgrowth of Neuro-2A cells in a dose-dependent manner. However, some of the recent studies have questioned its agonist nature and have suggested that it acts as a positive modulator of ligand-Induced GFRα/RET signaling and prolongs the GDNF-induced RET phosphorylation. Shown to be effective in treating small fiber neuropathy caused by non-myelinating Schwann cell dysfunction in diabetic murine models leading to improved thermononiception. Does not appear to be toxic up to ~100 µM.
A cell-permeable chlorostyrylquinolinylbutylamino compound that was shown to competitively bind to glial cell line-derived neurotrophic factor (GDNF) family receptor-α1 (GFRα1) and act as an agonist (IC50 = 10.4 µM for 125I-GDNF binding in Neuro-2A cells) and induce autophosphorylation of RET protein. Elicits neurite outgrowth of Neuro-2A cells in a dose-dependent manner. However, some of the recent studies have questioned its agonist nature and have suggested that it acts as a positive modulator of ligand-Induced GFRα/RET signaling and prolongs the GDNF-induced RET phosphorylation. Shown to be effective in treating small fiber neuropathy caused by non-myelinating Schwann cell dysfunction in diabetic murine models leading to improved thermononiception. Does not appear to be toxic up to ~100 µM.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GFRα1
Reversible: yes
Target IC50: 10.4 µM for ¹²⁵I-GDNF binding in Neuro-2A cells

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Hedstrom, K.L., et al. 2014. Proc. Natl. Acad. Sci. USA111, 2325.
Tokugawa, K., et al. 2003. Neurochem. Int.42, 81.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service