Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30510

Sigma-Aldrich

OxPhos Inhibitor, VLX600

Synonym(s):

OxPhos Inhibitor, VLX600, 1-(2-Pyridinyl)ethanone(6-methyl-5H-[1,2,4]triazino[5,6-b]indol-3-yl)hydrazone, ( Z)-6-Methyl-3-(2-(1-(pyridin-2-yl)ethylidene)hydrazinyl)-5H-[1,2,4]triazino[5,6-b]indole, 6-Methyl-3-((2 Z)-2-(1-(2-pyridinyl)ethylidene)hydrazino)-5H-[1,2,4]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15N7
Molecular Weight:
317.35
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark yellow

solubility

DMSO: 25 mg/mL

storage temp.

−20°C

SMILES string

CC1=C2C(=CC=C1)C3=C(N2)N=C(N=N3)NN=C(C)C4=CC=CC=N4

General description

A cell-permeable, bioavailable triazinoindolyl-hydrazone compound that displays preferential cytotoxicity towards quiescent cells in colon cancer 3-D microtissues (~ 6 µM). Inhibits the proliferation of HCT116 and several other human colon carcinoma and mouse cell lines in monolayer culture. Causes a significant reduction in mitochondrial oxidative phosphorylation in tumor cells thereby lowering their ATP levels and limiting their ability to respond to metabolically compromised microenvironment. Shown to increase AMP kinase phosphorylation and inhibit the phosphorylation of 4EBP1 and p70S6K, two downstream targets of mTOR. Induces LC-3II formation and autophagy in HCT 116, HCT116 HIF-1α -/-, and HT29 cells in a dose-dependent manner. Its sensitivity in HCT116 cells is enhanced under conditions of glucose starvation. Also reduces the growth of HCT116 and HT29 colon cancer xenografts in NMRI mice (~16 mg/kg, i.v. b.i.d, 5 d) and enhances oxaliplatin, irinotecan and 5-fluorouracil chemosensitivity. Displays attractive PK profile (t1/2 ~ 4 to 5 h).
A cell-permeable, bioavailable triazinoindolyl-hydrazone compound that displays preferential cytotoxicity towards quiescent cells in colon cancer 3-D microtissues (~ 6 µM). Inhibits the proliferation of HCT116 and several other human colon carcinoma and mouse cell lines in monolayer culture. Causes a significant reduction in mitochondrial oxidative phosphorylation in tumor cells thereby lowering their ATP levels and limiting their ability to respond to metabolically compromised microenvironment. Shown to increase AMP kinase phosphorylation and inhibit the phosphorylation of 4EBP1 and p70S6K, two downstream targets of mTOR. Induces LC-3II formation and autophagy in HCT 116, HCT116 HIF-1α -/-, and HT29 cells in a dose-dependent manner. Its sensitivity in HCT116 cells is enhanced under conditions of glucose starvation. Also reduces the growth of HCT116 and HT29 colon cancer xenografts in NMRI mice (~16 mg/kg, i.v. b.i.d, 5 d) and enhances oxaliplatin, irinotecan and 5-fluorouracil chemosensitivity. Displays attractive PK profile (t1/2 ~ 4 to 5 h).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
mitochondria

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Zhang, X., et al. 2014. Nat. Comm.5, 3295.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service