Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30341

Sigma-Aldrich

RPN13 Inhibitor, RA190

Synonym(s):

RPN13 Inhibitor, RA190, 3,5- bis(3,4-Dichlorobenzylidene)-1-( S-2-amino-3-phenyl)-piperidin-4-one, HCl, ADRM1 Inhibitor, Adhesion Regulating Molecule 1 Inhibitor, ARM1 Inhibitor; GP110 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H22Cl4N2O2 · xHCl
CAS Number:
Molecular Weight:
560.30 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

water: 5 mg/mL
DMSO: 50 mg/mL

storage temp.

−20°C

SMILES string

C1C(=CC2=CC(=C(C=C2)Cl)Cl)C(=O)C(=CC3=CC(=C(C=C3)Cl)Cl)CN1C(=O)C(CC4=CC=CC=C4)N.Cl

InChI

1S/C28H22Cl4N2O2.ClH/c29-22-8-6-18(12-24(22)31)10-20-15-34(28(36)26(33)14-17-4-2-1-3-5-17)16-21(27(20)35)11-19-7-9-23(30)25(32)13-19;/h1-13,26H,14-16,33H2;1H

General description

A cell-permeable bis-benzylidine piperidone that covalently modifies RPN13/ADRM1 via Michael addition between its electrophilic enone and the nucleophilic RPN13 Cys88, most likely freeing pleckstrin-like receptor for ubiquitin (Pru) domain from intramolecular interaction with the UCH37-binding domain and thereby allowing non-proteasome-associated RPN13 to compete against proteasome 19S regulatory particle- (RP-) incorporated RPN13 for ubiquitinated substrates binding, without affecting RPN13 RP assembly, 20S core particle (CP) proteolytic activities, or the deubiquitinase activity of UCH37 or RP. RPN13 blockage not only prevents many pro-apoptotic factors from degradation, the buildup of cellular ubiquitinated proteins in general also effectively triggers ER unfold protein response (UPR), accounting for the observed RA190 anticancer activity, notably in Multiple Myeloma (IC50 in nM = 35/MM.1S, 50/NCI-H929, 75/ANBL6,100/RPMI-8226), HPV+ cervical cancer (IC50 in nM = 150/HeLa, 300/CasKi, 750/HiHa), and HPV16-transformed oral keratinocyte HOK-16B (IC50 = 600 nM) cultures. RA190 is bioavailable via either oral or intraperitoneal administration in mice with tissue distribution in blood and major organs except brain and effectively inhibits NCI-H929 (20 mg/kg/d, i.p.), ES-2 (10 mg/kg/d, i.p.), and TC-1 (40 mg/kg/72 h, p.o.) tumor growths, as well as prevents 4UbFL reporter degradation in skin via topical application in mice in vivo without signs of adverse effects to animals or compromised host immune response. Synergizes with the 20S CP proteolytic activity inhibitor Bortezomib (Cat. No. 504314) in HeLa killing and is active against Bortezomib-resistant RPMI-8226-V10R and ANBL6-V10R cultures.
A cell-permeable, bioavailable, non-toxic bis-benzylidine piperidone compound that directly and covalently binds to Cys88 of ubiquitin receptor RPN13 Pru domain (pleckstrin-like receptor for ubiquitin) in the 19S regulatory particle and inhibits proteasome function leading to rapid accumulation of polyubiquitinated proteins. Does not affect the chymotryptic, tryptic, and peptidyl-glutamyl peptide-hydrolyzing activities of the 20S core particle. Exhibits anti-proliferative effects against multiple myeloma cells (IC50 = 100 nM) and HPV transformed cells (IC50 = 300 nM). Acts synergistically with bortezomib (Cat. No. 504314) to diminish cervical cancer cell viability and is effective even in multiple myeloma cells that are resistant to bortezomib treatment. Elevates p53 and p53-regulated genes in HeLa, CaSki, and SiHa cervical cancer cells as observed with bortezomib. Shown to induce apoptosis via induction of endoplasmic reticulum stress and up-regulate ATF-4 protein and CHOP-10 and XBP1s mRNA levels prior to apoptosis in MM.1S and HeLa cells. Reduces the growth of multiple myeloma and ovarian cancer xenografts in mice without affecting spontaneous HPV-specific CD8+ T cell responses. Displays desirable pharmacokinetic properties with Cmax in about 2 hours and t1/2α = 4.2 h; t1/2β = 25.5 h following a 10 mg/kg dose (i.p).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
RPN13 Inhibitor, RA190, is a cell-permeable compound that binds to cysteine 88 of ubiquitin receptor RPN13 Pru domain in the 19S regulatory particle and inhibits proteasome function.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
RPN13/ADRM1
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution

Other Notes

Anchoori, R.K., et al. 2013. Cancer Cell.24, 791.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service