Skip to Content
MilliporeSigma
All Photos(1)

Documents

529543

Millipore

Potassium Acetate, Molecular Biology Grade

Synonym(s):

Potassium Acetate, Molecular Biology Grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3KO2
CAS Number:
Molecular Weight:
98.14
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.25

grade

Molecular Biology

Quality Level

Assay

≥99% dry basis (perchloric acid titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: soluble

cation traces

heavy metals (as Pb): ≤0.0004%

foreign activity

DNases, proteases, RNases, none detected

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C2H4O2.K/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

SCVFZCLFOSHCOH-UHFFFAOYSA-M

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service