Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

528245

Sigma-Aldrich

PLD Inhibitor, FIPI

The PLD Inhibitor, FIPI controls the biological activity of PLD. This small molecule/inhibitor is primarily used for Membrane applications.

Synonym(s):

PLD Inhibitor, FIPI, Compound 4k, PLD Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$224.00

$224.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$224.00

About This Item

Empirical Formula (Hill Notation):
C23H24FN5O2
CAS Number:
Molecular Weight:
421.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$224.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(C1=CC2=C(C=CC(F)=C2)N1)NCCN3CCC(N4C(NC5=C4C=CC=C5)=O)CC3

General description

A cell-permeable, PLD1/2 inhibitor (IC50 = 1 nM and 10 nM for PLD1 and PLD2, respectively, in a CHO cell based assay, and IC50 = 20 nM for PLD2 in a biochemical assay), that displays no activity toward MitoPLD, and is selective for PLD1 and PLD2 over other PLD superfamily enzymes. It is also shown to block PLD2-dependent effects on cytoskeletal reorganization and cell trafficking in CHO cells, and diminish fMLP-directed neutrophil chemotaxis in HL60 cells at 750 nM. Unlike 1-butanol, this compound does not affect glucose-stimulated insulin secretion in Min6 pancreatic β-cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Su, W., et al. 2009. Mol Pharmacol75, 437.
Monovich, L., et al. 2007. Bioorg Med Chem17, 2310.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruth Ndathe et al.
Frontiers in plant science, 15, 1356699-1356699 (2024-04-29)
Core protein components of the abscisic acid (ABA) signaling network, pyrabactin resistance (PYR), protein phosphatases 2C (PP2C), and SNF1-related protein kinase 2 (SnRK2) are involved in the regulation of stomatal closure and gene expression downstream responses in Arabidopsis thaliana. Phosphatidic
Syed S Hussain et al.
Cell reports, 36(4), 109451-109451 (2021-07-29)
Lipid droplets (LDs) are dynamic organelles that undergo dynamic changes in response to changing cellular conditions. During nutrient depletion, LD numbers increase to protect cells against toxic fatty acids generated through autophagy and provide fuel for beta-oxidation. However, the precise

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service