Skip to Content
MilliporeSigma
All Photos(2)

Documents

528201

Sigma-Aldrich

PNU-120596

A urea derivative that acts as a potent, selective, and reversible positive allosteric modulator for the α7 subtype of neural nicotinic acetylcholine receptors

Synonym(s):

PNU-120596, α7 Neuronal Nicotinic Acetylcholine Receptor Agonist, PNU-120596, 1-(5-chloro-2,4-dimethoxy-phenyl)-3-(5-methyl-isoxazol-3-yl)-urea, a7 nAChR agonist, PNU-120596

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14ClN3O4
CAS Number:
Molecular Weight:
311.72
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C13H14ClN3O4/c1-7-4-12(17-21-7)16-13(18)15-9-5-8(14)10(19-2)6-11(9)20-3/h4-6H,1-3H3,(H2,15,16,17,18)

InChI key

CEIIEALEIHQDBX-UHFFFAOYSA-N

General description

A urea derivative that acts as a potent, selective, and reversible positive allosteric modulator for the α7 subtype of neural nicotinic acetylcholine receptors (EC50 = 216 nM), with no significant effect on α4β2, α3β4 and α9α10 receptors. Specifically increase a-7 nAChR-mediated channel currents and channel mean open time in cultured neurons (~ 1 mM). However, it does neither affect ion selectivity nor unitary conductance. Shown to cross the blood-brain barrier. Causes conformational changes in the ligand binding domain of a7 nicotinic receptors that partially overlap with those caused by acetylcholine. Causes changes in cysteine accessibility at the inner beta sheet, transition zone, and agonist binding site. Also reported to potentiate agonist-evoked GABAergic postsynaptic inward currents in somatodendritic membrane of hippocampal interneurons.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hurst, R., et al. 2005. J. Neurosci. 25, 4396.
Barron, S., et al. 2009. Mol. Pharm.76, 253.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service