Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

528121

Sigma-Aldrich

PI 3-K/PDK-1 Inhibitor, NVP-BAG956

The PI 3-K/PDK-1 Inhibitor, NVP-BAG956, also referenced under CAS 853910-02-8, controls the biological activity of PI 3-K/PDK-1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PI 3-K/PDK-1 Inhibitor, NVP-BAG956, α,α,-Dimethyl-4-(2-methyl-8-(2-(3-pyridinyl)ethynyl)-1H-imidazo[4,5-c]quinolin-1-yl)-benzeneacetonitrile, BAG956, PI 3-K Inhibitor XVI, PDK1 Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H21N5
CAS Number:
Molecular Weight:
427.50
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C28H21N5/c1-19-32-26-17-31-25-13-8-20(6-7-21-5-4-14-30-16-21)15-24(25)27(26)33(19)23-11-9-22(10-12-23)28(2,3)18-29/h4-5,8-17H,1-3H3

InChI key

GVPAGJWVBUZHNQ-UHFFFAOYSA-N

General description

A cell-permeable imidazoquinoline compound that acts as a potent, reversible, and ATP-competitive PI 3-K/PDK1 dual kinase inhibitor (IC50 = 56, 446, 35, 117, and 245 nM against p110α, p110β, p110δ, p110γ, and PDK1, respectively), displaying much reduced potency against VEGFR1 (IC50 = 2.56 µM) and little or no activity toward a panel of 15 other kinases (IC50 >10 µM). Shown to effectively inhibit the constitutive phosphorylation of PKB (T308 & S473; IC50 ≤45 nM) as well as PKB downstream effectors GSK3β (S9), FKHRL1 (T32), and p70S6K (T389) in human glioma U87MG and adeonocarcinoma PC3M cultures with PTEN deletion in vitro and retard PC3M-derived tumor growth in mice (by 50%; 75 mg/kg; twice per day p.o.) in vivo. Also reported to exhibit antiproliferative activities against BCRF-ABL- and mutant FLT3-expressing cells either alone or in combination with other PI3K/AKT, mTOR, and tyrosine kinase inhibitors, LY294002 (Cat. Nos. 440202 & 440204), Rapamycin (Cat. No. 553210), and PKC412 (Cat. No. 539648).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Marone, R., et al. 2009. Mol. Cancer Res.7, 601.
Weisberg, E., et al. 2008. Blood111, 3723.
Stauffer, F., et al. 2008. Bioorg. Med. Chem. Lett.18, 1027.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service