Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

528120

Sigma-Aldrich

PIPER - CAS 213416-70-7 - Calbiochem

A perylene-based ligand that potently inhibits human telomerase by binding to G-quadruplex DNA.

Synonym(s):

PIPER - CAS 213416-70-7 - Calbiochem, N,Nʹ- bis[2-(1-Piperidino)ethyl]-3,4,9,10-perylenetetracarboxylic Diimide, Telomerase Inhibitor IV, N,Nʹ-bis[2-(1-Piperidino)ethyl]-3,4,9,10-perylenetetracarboxylic Diimide, Telomerase Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H36N4O4
CAS Number:
Molecular Weight:
612.72
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark purple

solubility

5% acetic acid: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A perylene-based ligand that potently inhibits human telomerase by binding to G-quadruplex DNA. The strongest binding site for PIPER appears to be the 3′-boundary of the G-quadruplex. Can also bind non-specifically to nucleic acids. May also be useful as an antiproliferative agent.
A perylene-based ligand that potently inhibits human telomerase by binding to G-quadruplex DNA. The strongest binding site for PIPER appears to be the 3′-boundary of the G-quadruplex. Can also bind non-specifically to nucleic acids. May be useful as an antiproliferative agent.

Biochem/physiol Actions

Cell permeable: no
Primary Target
human telomerase
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Han, H., et al. 1999. Biochemistry38, 6981.
Fedoroff, O.Y., et al. 1998. Biochemistry 37, 12367.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service