Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

527460

Sigma-Aldrich

PhosphoDetect Anti-PKR (pThr⁴⁵¹) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Double-stranded RNA-dependent Protein Kinase, Phospho-Specific (Thr⁴⁵¹)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

phosphorylation (pThr451)

Gene Information

human ... EIF2AK2(5610)

General description

Note: 1 T = 1 test. Sufficient for 10 Western miniblots.
Rabbit polyclonal antibody adsorbed against non-phosphopeptide corresponding to the immunogen phophorylation site, followed by immunoaffinity chromatography. Recognizes the ~65-68 kDa PKR protein phosphorylated at Thr451.
Recognizes the ~65-68 kDa PKR protein phosphorylated at Thr451.
This PhosphoDetect Anti-PKR (pThr⁴⁵¹) Rabbit pAb is validated for use in Immunoblotting, Immunohistochemistry for the detection of PKR (pThr⁴⁵¹).

Immunogen

a synthetic phosphopeptide corresponding to amino acids surrounding the Thr⁴⁵¹ phosphorylation site of human PKR (double-stranded RNA-dependent protein kinase)

Application

Immunoblotting (1:1000)

Immunohistochemistry (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

In Dulbecco′s PBS (without Mg+2 or Ca+2), 0.1% BSA, 50% glycerol, pH 7.3.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Analysis Note

Positive Control
IFNγ-stimulated HeLa cells.

Other Notes

Cuddihy, A.R., et al.1999. Oncogene8, 2690.
Srivastava, S.P., et al. 1998. J. Biol. Chem.273, 2416.
Clemens, M.J. 1997. Int. J. Biochem. Cell Biol.29, 945.
When used at a 1:1000 dilution there is sufficient antibody for 10 blots at 10 ml/blot. This antibody has also been reported to work for immunohistochemistry. Variables associated with assay conditions will dictate the optimal dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Angel García et al.
PloS one, 6(8), e23887-e23887 (2011-09-03)
The chemotherapeutic drug 5-FU is widely used in the treatment of a range of cancers, but resistance to the drug remains a major clinical problem. Since defects in the mediators of apoptosis may account for chemo-resistance, the identification of new
Sharmistha Sarkar et al.
Viruses, 16(7) (2024-07-27)
Negative-strand RNA viruses form cytoplasmic inclusion bodies (IBs) representing virus replication foci through phase separation or biomolecular condensation of viral and cellular proteins, as a hallmark of their infection. Alternatively, mammalian cells form stalled mRNA containing antiviral stress granules (SGs)
Nishi R Sharma et al.
PLoS pathogens, 13(10), e1006677-e1006677 (2017-10-31)
TIA-1 positive stress granules (SG) represent the storage sites of stalled mRNAs and are often associated with the cellular antiviral response. In this report, we provide evidence that Kaposi's sarcoma-associated herpesvirus (KSHV) overcomes the host antiviral response by inhibition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service