Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

526528

Sigma-Aldrich

PIM-Kinase Inhibitor IX, SGI-1776

The PIM-Kinase Inhibitor IX, SGI-1776 controls the biological activity of PIM-Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PIM-Kinase Inhibitor IX, SGI-1776, N-((1-Methylpiperidin-4-yl)methyl)-3-(3-(trifluoromethoxy)phenyl)imidazo[1,2-b]pyridazin-6-amine, 2H₂SO₄

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22F3N5O · 2H2SO4
CAS Number:
Molecular Weight:
601.57
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

DMSO: 100 mg/mL, clear, nearly colorless
water: 100 mg/mL, clear, nearly colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

CN(CC1)CCC1CNC(C=C2)=NN3C2=NC=C3C4=CC(OC(F)(F)F)=CC=C4

General description

A cell-permeable imidazopyridazine compound that acts as a potent, reversible and ATP-competitive inhibitor of Pim-1,2,3, Flt-3 and haspin kinase activities (IC50 = 7, 363, 69, 44 and 34 nM, respectively) with moderate (40% inhibition at 1 µM for c-kit) to excellent selectivity over CDKs, Aurora, Chk1, IKK, JNK, Abl, Raf, PKA, PKC and PI 3-K among 300-kinases. Shown to inhibit RNA synthesis, block phosphorylation of c-Myc-Ser62 and STAT3-Tyr705, and decrease c-Myc and Mcl-1 protein levels; cause cell cycle arrest and induce apoptosis in CLL lymphocytes and in AML cells, and efficiently arrest the growth of several prostate cancer cells (IC50<10 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstituion, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Chen, L.S., et al. 2011. Blood118, 693.
Chang, M., et al. 2010. Mol. Cancer Ther.9, 2478.
Mumenthaler, S.M., et al. 2009. Mol. Cancer Ther.8, 2882.
Chen, L.S., et al. 2009. Blood 114, 4150.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service