Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

525330

Sigma-Aldrich

PFKFB3 Inhibitor, 3PO

PFKFB3 Inhibitor, 3PO, is a cell-permeable, selective inhibitor of PFK-2 (6-phosphofructo-2-Kinase) activity.

Synonym(s):

PFKFB3 Inhibitor, 3PO, (2 E)-3-Pyridin-3-yl-1-pyridin-4-ylprop-2-en-1-one, 3-(3-Pyridinyl)-1-(4-pyridinyl)-2-propen-1-one, 6-Phosphofructo-2-kinase/fructose-2,6-bisphosphatase 3 Inhibitor, 3PO

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$181.00

$181.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$181.00

About This Item

Empirical Formula (Hill Notation):
C13H10N2O
Molecular Weight:
210.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

$181.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

n1cc(ccc1)\C=C\C(=O)c2ccncc2

InChI

1S/C13H10N2O/c16-13(12-5-8-14-9-6-12)4-3-11-2-1-7-15-10-11/h1-10H/b4-3+

InChI key

UOWGYMNWMDNSTL-ONEGZZNKSA-N

General description

A cell-permeable dipyridinyl-propenone compound that selectively blocks PFK-2 (6-phosphofructo-2-kinase) activity of PFKFB3 isozyme over PFK-1(6-phosphofructo-1-kinase) in a mixed-mode manner. Rapidly decreases fructose-2,6-bisphosphate (F-2,6-P2) and 2-deoxyglucose uptake, and lactate secretion; efficiently arrests proliferation of transformed cells (IC50 ~ 1.4 µM in NHBE-ht/LT/ras) and suppresses tumor growth in several xenograft mouse models (70 mg/kg, i.p.). The cytostatic and cytotoxic effects of 3PO is shown to be highly sensitive to the intracellular levels of F-6-P (fructose-6-phosphate) and F-2,6-P2.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 6 months at -70°C.

Other Notes

Clem, B., et al. 2008. Mol. Cancer Ther.7, 110.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian Clem et al.
Molecular cancer therapeutics, 7(1), 110-120 (2008-01-19)
6-phosphofructo-1-kinase, a rate-limiting enzyme of glycolysis, is activated in neoplastic cells by fructose-2,6-bisphosphate (Fru-2,6-BP), a product of four 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase isozymes (PFKFB1-4). The inducible PFKFB3 isozyme is constitutively expressed by neoplastic cells and required for the high glycolytic rate and anchorage-independent
Juliana Escher Toller-Kawahisa et al.
Nature communications, 14(1), 4280-4280 (2023-07-18)
Neutrophils rely predominantly on glycolytic metabolism for their biological functions, including reactive oxygen species (ROS) production. Although pyruvate kinase M2 (PKM2) is a glycolytic enzyme known to be involved in metabolic reprogramming and gene transcription in many immune cell types
Gangqi Wang et al.
Arteriosclerosis, thrombosis, and vascular biology, 40(2), 350-364 (2019-12-13)
Endothelial cells exposed to laminar shear stress express a thick glycocalyx on their surface that plays an important role in reducing vascular permeability and endothelial anti-inflammatory, antithrombotic, and antiangiogenic properties. Production and maintenance of this glycocalyx layer is dependent on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service