Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

525186

Sigma-Aldrich

Phospholipase C, Bacillus cereus

Synonym(s):

Phospholipase C, Bacillus cereus, PLC

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54
Pricing and availability is not currently available.

description

RTECS - SZ6116000

Quality Level

form

solid

specific activity

≥50 units/mg dry wt.

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to brownish

solubility

aqueous buffer: soluble
water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Native phospholipase C from Bacillus cereus. Catalyzes the hydrolysis of lecithins to 1,2-diglycerides and choline phosphate.

Warning

Toxicity: Harmful (C)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1 µmol of lecithin per min at 37°C, pH 8.0.

Other Notes

Rahman, S.M., et al. 1995. Infect. Immun. 63, 259.
Bhamidipati, S.P., and Hamilton, J.A. 1993. J. Biol. Chem. 268, 2431.
Nieva, J.L., et al. 1993. Biochemistry 32, 1054.
Hendrickson, H.S., et al. 1992. Biochemistry 31, 12169.
Slack, B.E., et al. 1991. J. Biol. Chem. 266, 24503.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service