Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

524750

Millipore

TBS Tablets

Dissolving one tablet in 500 ml of deionized H2O yields 150 mM NaCl, 50 mM Tris-HCl buffer, pH 7.6 at 25°C. Supplied in convenient blister packs. Note: 1 each = 10 tablets.

Synonym(s):

Tris Buffered Saline 10 Tablets

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$98.80

$98.80


Available to ship onMarch 31, 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$98.80

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

$98.80


Available to ship onMarch 31, 2025Details


Request a Bulk Order

Quality Level

packaging

pkg of 10 tablets

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

15-25°C

General description

Dissolving one tablet in 500 ml of deionized H2O yields 150 mM NaCl, 50 mM Tris-HCl buffer, pH 7.6 at 25°C. Supplied in convenient blister packs. Note: 1 each = 10 tablets.
Tris-buffered saline supplied as convenient blister packs. Note: 1EA = 10 tablets.

Warning

Toxicity: Irritant (B)

Preparation Note

Dissolve one tablet in 500 ml dH₂O to yield 150 mM NaCl, 50 mM Tris-HCl buffer, pH 7.6 at 25°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service