Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

524715

Sigma-Aldrich

Phosphodiesterase, Type V, cGMP-Specific, Bovine, Recombinant, Spodoptera frugiperda

Synonym(s):

PDE V, Phosphodiesterase V, cGMP-binding cGMP-specific PDE

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
$471.00

$471.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μL
$471.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$471.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

Quality Level

specific activity

≥50,000 units/mL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, bovine c-GMP-specific phosphodiesterase expressed in S. frugiperda insect cells. Supplied as partially purified, catalytically active, dimeric holoenzyme. A target enzyme for Viagra (sildenafil). Suitable for enzymatic studies and as a standard in Western blots using Anti-Phosphodiesterase V Antibody (Cat. No. 524583).
Recombinant, bovine c-GMP-specific phosphodiesterase expressed in S. frugiperda insect cells. Supplied as partially purified, catalytically active, dimeric holoenzyme. A target enzyme for Viagra (sildenafil). Suitable for enzymatic studies and as a standard in Western blots using Anti-Phosphodiesterase V Antibody (Cat. No. 524583). For enzymatic studies a dilution of 1:1000 recommended. For use as a immunoblot standard 2 to 3 µl per lane is recommended. In immunoblotting, gives band at ~92 kDa.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 pmol cGMP per min at 25°C, pH 7.2.

Physical form

In sodium phosphate buffer, 20% glycerol, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Gibson, L.C.D., et al. 2006. Eur. J. Pharmacol.538, 39.
Webb, D.J., et al. 1999. Am. J. Cardiol. 83, 21C.
Turko, I.V., et al. 1998. J. Biol. Chem. 273, 6460.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service