Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

523310

Sigma-Aldrich

Pterostilbene, Pterocarpus marsupium

A cell-permeable methoxylated analog of Resveratrol that displays antioxidant, antiproliferative, and hypoglycemic properties.

Synonym(s):

Pterostilbene, Pterocarpus marsupium, trans-3,5-Dimethoxy-4ʹ-hydroxystilbene

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$268.00

$268.00


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
$268.00

About This Item

Empirical Formula (Hill Notation):
C16H16O3
CAS Number:
Molecular Weight:
256.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$268.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL
methanol: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O(C)c1cc(cc(c1)\C=C\c2ccc(cc2)O)OC

InChI

1S/C16H16O3/c1-18-15-9-13(10-16(11-15)19-2)4-3-12-5-7-14(17)8-6-12/h3-11,17H,1-2H3/b4-3+

InChI key

VLEUZFDZJKSGMX-ONEGZZNKSA-N

General description

A cell-permeable methoxylated analog of Resveratrol (Cat. No. 554325) that displays antioxidant, antiproliferative, and hypoglycemic properties. Moderately inhibits COX-1 and -2 activities (IC50 = 19.8 µM and 83.9 µM) respectively, induces apoptosis in HL60 cells (IC50 = 70 µM), prevents DMBA-induced preneoplastic lesions (ED50 = 4.8 µM) and decreases plasma glucose level in streptozotocin-induced diabetic rats comparable to that of Metformin.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
COX-1 and COX-2 activities
Product does not compete with ATP.
Reversible: no
Target IC50: 19.8 µM, 83.9 µM against COX-1 and -2 activities, respectively; 70 µM for inducing apoptosis in HL60 cells; ED50 = 4.8 µM for preventing DMBA-induced preneoplastic lesions

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C.). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Roberti, M., et al. 2003. J. Med. Chem.46, 3546.
Rimando, A.M., et al. 2002. J. Agric. Food Chem.50, 3453.
Manickam, M., et al. 1997. J. Nat. Prod.60, 609.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service