Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.09230

Sigma-Aldrich

ER Export Inhibitor II, FLI-06

Synonym(s):

ER Export Inhibitor II, FLI-06, Cyclohexyl 2,7,7-trimethyl-4-(4-nitrophenyl)-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carboxylate, Cyclohexyl 4-(4ʹ-nitrophenyl)-2,7,7-trimethyl-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carboxylate, FLI06

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N2O5
Molecular Weight:
438.52
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, non-toxic, dihydropyridine compound that prevents cellular maturation/glycosylation and surface expression of Notch, APP, Klotho (Effect. conc. = 10 µM in HEK293, HeLa, U2OS cultures) in a rapid and reversible manner by blocking events prior to cargo assembly at the endoplasmic reticulum exit sites (ERES) without affecting cytoskeleton, endocytosis, or other post-ERES events targeted by BFA, GCA, Exo1, A5, Retro-2, U18666A, or Vacuolin-1 (Cat. nos. 203729, 345862, 341220, 444805, 554715, 662015, & 673000, respectively). Notch signaling is shown to be effectively inhibited by treatment with either FLI-06 or the γ-Secretase inhibitior DAPT (Cat. nos. 565770 & 565784) in both C2C12 cultures (10 µM FLI-06) in vitro and in zebrafish embryos (50 µM FLI-06) in vivo.
A cell-permeable, non-toxic, dihydropyrimidine compound that prevents cellular glycosylation and surface expression of Notch, APP, Klotho (Effect. conc. = 10 µM in HEK293, HeLa, U2OS cultures) in a rapid and reversible manner by blocking events prior to cargo assembly at the ER exit sites (ERES) without affecting other post-ERES events targeted by BFA, GCA, Exo1, A5, Retro-2, U18666A, or Vacuolin-1 (Cat. nos. 203729, 345862, 341220, 444805, 554715, 662015, & 673000, respectively). Notch signaling is shown to be effectively inhibited by treatment with either FLI-06 or the γ-Secretase inhibitior DAPT (Cat. nos. 565770 & 565784) in both C2C12 cultures (10 µM FLI-06) in vitro and in zebrafish embryos (50 µM FLI-06) in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ER
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Kramer, A., et al. 2013. Nat. Chem. Biol.9, 731.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service