Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5091RU0

Sigma-Aldrich

Human Metapneumovirus DFA Reagent, RUO

Synonym(s):

hMPV, Human metapneumovirus

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$323.00

$323.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 ML
$323.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.41

$323.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

clone

monoclonal

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

immunofluorescence: suitable

isotype

IgG

shipped in

wet ice

General description

Diagnostics Kit
Ready to use purified anti-hMPV FITC conjugated monoclonal antibodies in solution. The reagent is offered in a 2 mL dropper vial of FITC-conjugated monoclonal antibodies specific for Human Metapneumovirus (HMPV) in 0.2% BSA in PBS, Tween, with Sodium Azide. Used for direct fluorescence assay (DFA) where stained specimens are identified using immunofluorecsence microscopy.

Specificity

Human Metapneumovirus

Application

Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

2 mL dropper vial of monoclonal antibodies specific for Human Metapneumovirus (HMPV) in 0.2% BSA in PBS, Tween, with Sodium Azide.
Format: Purified

Storage and Stability

WARNING: Sodium azide, present in this reagent can form potentially explosive metal azides with lead and copper pipes. Upon disposal, flush with large volumes of water to prevent build-up in plumbing.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prospective study of human metapneumovirus detection in clinical samples by use of light diagnostics direct immunofluorescence reagent and real-time PCR.
Landry, Marie L, et al.
Journal of Clinical Microbiology, 46, 1098-1100 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service