Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.08838

Millipore

Puromycin, Dihydrochloride, Sterile-Filtered Aqueous Solution, Cell Culture-Tested

Synonym(s):

Puromycin, Dihydrochloride, Sterile-Filtered Aqueous Solution, Cell Culture-Tested, (2 S)-2-Amino-N-((2 S,3 S,4 R,5 R)-5-(6-(dimethylamino)purin-9-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl)-3-(4-methoxyphenyl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29N7O5 · 2HCl
CAS Number:
Molecular Weight:
544.43
MDL number:
UNSPSC Code:
12352101

Assay

≥98% (HPLC)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

impurities

≤10 EU/mg Endotoxin (dry weight)

storage temp.

−20°C

InChI

1S/C22H29N7O5.2ClH/c1-28(2)19-17-20(25-10-24-19)29(11-26-17)22-18(31)16(15(9-30)34-22)27-21(32)14(23)8-12-4-6-13(33-3)7-5-12;;/h4-7,10-11,14-16,18,22,30-31H,8-9,23H2,1-3H3,(H,27,32);2*1H/t14-,15+,16?,18+,22+;;/m0../s1

InChI key

MKSVFGKWZLUTTO-USYAMCSGSA-N

General description

An aminonucleoside antibiotic that acts as a prokaryotic and eukaryotic protein synthesis inhibitor.

Biochem/physiol Actions

Primary Target
RNA
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied at 10 mg/ml in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.

Other Notes

Chow, S.C., et al. 1995. Exp. Cell Res.216, 149.
Kaufman, S.H., et al. 1993. Cancer Res.53, 3976.
de la Luna, S. and Ortin, J. 1992. Methods Enzymol.216, 376.
Chow, S.C., et al. 1991. Exp. Cell Res.216, 149.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service