Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.08509

Sigma-Aldrich

NS309

Synonym(s):

NS309, Ca2+ -activated K+ Channels Activator, NS309, NS-309, NS 309

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$106.00

$106.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$106.00

About This Item

Empirical Formula (Hill Notation):
C8H4Cl2N2O2
CAS Number:
Molecular Weight:
231.04
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$106.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mM

storage temp.

−20°C

SMILES string

Clc1[c]2[c](ccc1Cl)=C(C(=O)N=2)NO

InChI

1S/C8H4Cl2N2O2/c9-4-2-1-3-6(5(4)10)11-8(13)7(3)12-14/h1-2,14H,(H,11,12,13)

InChI key

CVOUSAVHMDXCKG-UHFFFAOYSA-N

General description

A potent activator of Ca2+ -activated K+ channels of SK and IK types but not BK type channels. NS309 is at least 1000 times more potent than 1-EBIO and at least 30 times more potent than DC-EBIO when the compounds are compared on the same cell. NS309 blocks L-type Calcium channels with IC50 = 10 µM/L. Recently, this compound has been shown to produce endothelium-dependent hyperpolarizations in guinea pig carodid arteries by opening KCa2.3 and KCa3.1 channels demonstrating a new therapeutic option for treating hypertension.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
Ca2+-activated K+ channels

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Leurangue, V. et al. 2008. Naunyn Schmiedebergs Arch. Pharmacol.377, 101.
Morimura, K. et al. 2006. J. Pharmacol. Sci.100, 237.
Strobaek, D. et al. 2004. Biochim. Biophys. Acta.1665, 1.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service