Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.08437

Sigma-Aldrich

Pepstatin A

InSolution, ≥98%, Synthetic

Synonym(s):

InSolution Pepstatin A, Synthetic, Isovaleryl-Val-Val-4-amino-3-hydroxy-6-methylheptanoyl-Ala-4-amino-3-hydroxy-6-methylheptanoic Acid, Pepstatin A, Synthetic InSolution, Pepstatin A, Synthetic InSolution, Isovaleryl-Val-Val-4-amino-3-hydroxy-6-methylheptanoyl-Ala-4-amino-3-hydroxy-6-methylheptanoic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H63N5O9
CAS Number:
Molecular Weight:
685.89
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

InChI

1S/C34H63N5O9/c1-17(2)12-23(37-33(47)31(21(9)10)39-34(48)30(20(7)8)38-27(42)14-19(5)6)25(40)15-28(43)35-22(11)32(46)36-24(13-18(3)4)26(41)16-29(44)45/h17-26,30-31,40-41H,12-16H2,1-11H3,(H,35,43)(H,36,46)(H,37,47)(H,38,42)(H,39,48)(H,44,45)/t22-,23?,24?,25?,26?,30-,31-/m0/s1

InChI key

FAXGPCHRFPCXOO-JHTFMCFMSA-N

Biochem/physiol Actions

Primary Target
aspartic proteases
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Iva-Val-Val-Sta-Ala-Sta

Physical form

A 25 mM (10 mg/583 µL) sterile-filtered solution of Pepstatin A, Synthetic (Cat. No. 516481) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 4 months at -20°C. Use only fresh DMSO for further dilutions.

Other Notes

Isahara, K., et al. 1999. Neuroscience91, 233.
Kanamori, S., et al. 1998. Biochem. Biophys. Res. Commun.251, 204.
Szewczuk, Z., et al. 1992. Int. J. Pept. Prot. Res.40, 233.

Carraway, R.E., et al. 1991. Peptides12, 107.
Eid, M., et al. 1981. Biochem. J.197, 465.
Umezawa, H., et al. 1970. J. Antibiot.23, 259.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service