Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.08150

Sigma-Aldrich

LY 344864 Hydrochloride

Synonym(s):

LY 344864 Hydrochloride, 5-HT 1F Serotonin Receptor Agonist, LY 344864, LY344864, LY-344864, 5-HT1F Serotonin Receptor Agonist, LY 344864, LY344864, LY-344864

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22FN3O · xHCl
CAS Number:
Molecular Weight:
351.42 (free base basis)
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 10 mM
DMSO: 100 mM

storage temp.

2-8°C

InChI

1S/C21H22FN3O.ClH/c1-25(2)16-8-10-20-18(12-16)17-11-15(7-9-19(17)24-20)23-21(26)13-3-5-14(22)6-4-13;/h3-7,9,11,16,24H,8,10,12H2,1-2H3,(H,23,26);1H/t16-;/m1./s1

InChI key

OKUHLSYESBLBCP-PKLMIRHRSA-N

General description

A selective, brain penetrant, full agonist for the 5-HT1F receptor (pKi = 8.2). It possesses little affinity for the 56 other serotonergic and non-serotonergic neuronal binding sites examined. When examined for its ability to inhibit forskolin-induced cyclic AMP accumulation in cells stably transfected with human 5-HT1F receptors, LY344864 was shown to be a full agonist producing an effect similar in magnitude to serotonin itself. Suitable for both in vitro and in vivo applications and its effects are often reversed with the 5-HT1B/1D antagonist GR 127935 (Cat. No. 508014)

Biochem/physiol Actions

Primary Target
5-HT1F

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Amrutkar, D. V. et al. 2012. Pain.153, 830.

Phebus, L. A. et al. 1997. Life Sci.61, 2117.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service