Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.06272

Sigma-Aldrich

SB 206553 Hydrochloride

Synonym(s):

SB 206553 Hydrochloride, 5-HT 2C Serotonin Receptor Antagonist, SB 206553, 5-HT 2B Serotonin Receptor Antagonist, SB 206553, SB206553, SB-206553

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$210.00

$210.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$210.00

About This Item

Empirical Formula (Hill Notation):
C17H16N4O · xHCl
CAS Number:
Molecular Weight:
292.34 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$210.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

3.2 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: soluble

storage temp.

2-8°C

SMILES string

[n]1(c2c(cc3c(c2)CCN3C(=O)Nc4cnccc4)cc1)C

InChI

1S/C17H16N4O/c1-20-7-4-12-10-16-13(9-15(12)20)5-8-21(16)17(22)19-14-3-2-6-18-11-14/h2-4,6-7,9-11H,5,8H2,1H3,(H,19,22)

InChI key

QJQORSLQNXDVGE-UHFFFAOYSA-N

General description

A selective 5-HT2C serotonin receptor inhibitor with lesser affinity for 5-HT2BR. Often used in conjunction with ketanserin to distinguish 5-HT2AR and 5-HT2CR activity. SB206553 has highest affinity for 5-HT2CR (Ki = 3.2nM) and less for 5-HT2BR (Ki = 5.5 nM) and 5-HT2AR (Ki = 2,300 nM).

Biochem/physiol Actions

Primary Target
5-HT2C
Secondary Target
5-HT2B

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ros, R. et al. 2013. PNAS.110, 348.

Knight, A. R. et al. 2004. Naunyn Schmiedebergs Arch. Pharmacol.370, 114.
Cussac, D. et al. 2002. Naunyn Schmiedebergs Arch. Pharmacol.365, 242.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service