Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.06238

Sigma-Aldrich

Citalopram Hydrobromide

Synonym(s):

Citalopram hydrobromide, 5-HT Transporter Inhibitor, Citalopram, Celexa, 1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21FN2O · HBr
CAS Number:
Molecular Weight:
405.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

1.8 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 10 mM
ethanol: 50 mM

storage temp.

2-8°C

SMILES string

Br[H].CN(C)CCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3

InChI

1S/C20H21FN2O.BrH/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;/h4-9,12H,3,10-11,14H2,1-2H3;1H

InChI key

WIHMBLDNRMIGDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A potent uptake inhibitor highly selective for 5-HT transporter without effect on noradrenalin or dopamine uptake (IC50 = 1.8, 8800 and 41000 nM for 5-HT, noradrenalin and dopamine, respectively). Clinically prescribed as an antidepressant drug. Widely used in research related to depressive disorders.

Biochem/physiol Actions

Potent and selective serotonin uptake inhibitor (Ki = 5.4 nM); antidepressant
Primary Target
5-HT Transporter

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Garriock, A., et al., 2010. Bio. Psychiatry.67, 133.
Gabriel, J.P., et al. 2009. J. Neurosci.29,10387.
Torres, G.E., et al. 2003. Nat. Rev. Neurosci.4, 13.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service