Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

506114

Sigma-Aldrich

p34cdk1/Cyclin B, Human, Recombinant, S. frugiperda

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

form

liquid

Quality Level

specific activity

≥20,000 U/mL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

foreign activity

Protease, phosphatase, none detected

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human p34Cdk1/cyclin B expressed in S. frugiperda insect cells. This kinase is a major regulator of the eukaryotic cell cycle. As a monomer, Cdk1 is inactive and requires association with cyclin B and phosphorylation of Thr161 to form an active complex. MW: 34000(p34Cdk1) and 55000(cyclinB).
Recombinant, human p34Cdk1/cyclin B expressed in S. frugiperda insect cells. This kinase is a major regulator of the eukaryotic cell cycle. As a monomer, p34cdc2 is inactive and requires association with cyclin B to form an active complex.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that catalyzes the transfer of 1 pmol of phosphate to histone H1 (10 µM) in 1 min at 30°C, pH 7.5.

Physical form

In 100 mM NaCl, 50 mM HEPES, 1 mM DTT, 100 µM EDTA, 50% glycerol, 0.01% BRIJ® 35 Detergent, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Kobayashi, H., et al. 1994. J. Biol. Chem. 269, 29153.
Atherton-Fessler, S., et al. 1993. Mol. Cell. Biol. 13, 1675.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service