Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.06071

Sigma-Aldrich

I-BET

InSolution, ≥98%, 50 mM in DMSO, A cell-permeable benzodiazepine compound

Synonym(s):

InSolution I-BET, ( S)-2-(6-(4-Chlorophenyl)-8-methoxy-1-methyl-4H-benzo[f][1,2,4]triazolo[4,3-a][1,4]diazepin-4-yl)-N-ethylacetamide, (S)-2-(6-(4-Chlorophenyl)-8-methoxy-1-methyl-4H-benzo[f][1,2,4]triazolo[4,3-a][1,4]diazepin-4-yl)-N-ethylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22ClN5O2
CAS Number:
Molecular Weight:
423.90
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

InChI

1S/C22H22ClN5O2/c1-4-24-20(29)12-18-22-27-26-13(2)28(22)19-10-9-16(30-3)11-17(19)21(25-18)14-5-7-15(23)8-6-14/h5-11,18H,4,12H2,1-3H3,(H,24,29)/t18-/m0/s1

InChI key

AAAQFGUYHFJNHI-SFHVURJKSA-N

General description

A cell-permeable benzodiazepine compound that binds the tandem bromodomains of BET (bromodomain and extra terminal domain) family members BRD2 (1-473), BRD3 (1-434), and BRD4 (1-477) with high affinity (Kd = 61.3, 50.5, and 55.2 nM, respectively, by ITC) and effectively competes against tetra-acetylated H4 peptide (Millipore cat. no. 12-379) for BRD2/3/4 binding (IC50 = 32.5, 42,4, and 36.1 nM, respectively, in competitive equilibrium binding assays), while exhibiting little affinity toward 5 other bromodomain-containing proteins (ATAD2, BAZ2B, CREBBP, PCAF, SP140) or a panel of 38 cellular enzymes, GPCRs, transporters, and ion channels. Shown to differentially modulate LPS-induced gene expression, notably suppressing LPS-induced upregulation of genes involved in inflammatory response, in murine BMDMs (bone marrow-derived macrophages) in vitro (30 min 1 µM pretreatment) and effectively prevent LPS-, heat-killed Salmonella typhimurium-, and CLP- (caecal ligation and puncture) induced death in mice in vivo (30 mg/kg i.v.). Cellular Brd2/3/4 knockdown using siRNA results in mostly the same effect as I-BET treatment in modulating LPS gene induction profile in murine BMDMs, but not without exceptions, indicating that not all BET-dependent transcription regulations are mediated via its binding to acetylated histones.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
tandem bromodomains of BET
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 50 mM (5 mg/236 µL) sterile-filtered solution of I-BET (Cat. No. 401010) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.

Other Notes

Nicodeme, E., et al. 2010. Nature468, 1119.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service