Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.06069

Sigma-Aldrich

3-Deazaneplanocin A hydrochloride

InSolution, ≥98%, 25 mM aqueous solution, EZH2 Inhibitor

Synonym(s):

InSolution EZH2 Inhibitor, DZNep, 3-Deazaneplanocin A, xHCl, yH₂O {x = 3 & y = 2}, (1S,2R,5R)-5-(4-Amino-1H-imidazo[4,5-c]pyridin-1-yl)-3-(hydroxymethyl)cyclopent-3-ene-1,2-diol, xHCl, yH₂O {x = 3 & y = 2}, NSC 617989

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O3 · 3HCl · 2H2O
CAS Number:
Molecular Weight:
407.68
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

storage temp.

−70°C

InChI

1S/C12H14N4O3/c13-12-9-7(1-2-14-12)16(5-15-9)8-3-6(4-17)10(18)11(8)19/h1-3,5,8,10-11,17-19H,4H2,(H2,13,14)/t8-,10-,11+/m1/s1

InChI key

OMKHWTRUYNAGFG-IEBDPFPHSA-N

General description

A cell-permeable compound that is shown (at 1 µM concentrations) to inhibit EZH2-mediated trimethylation of K27 on histone H3 and induces the expression of cell-cycle regulatory genes, p21 and p27, as well as the cell death regulator, FBXO32, in OCI-AML3 and HL-60 cells, whereby treatment with inhibitor increases p16 levels in the former, but not the latter of the two cultures. At concentrations between 200 nm and 2000 nM, this compound is found to dose-dependently deplete the expression of polycomb group proteins EZH2, SUZ12, and EED in cultured and primary AML cell extracts. At concentrations up to ≥ 1000 nM, DZNep dose-dependently increases the percentage of apoptotic cells up to > ~ 38%, with greater potency against OCI-AML3 than HL-60 cultures and inhibits colony growth up to > ~ 85% for both cell lines. In OCI-AML3 cultures, 1000 nM of treatment demonstrates a significant increase in the accumulation of cells in the G0/G1 phase (58.5%) with a concomitant decrease in the number of cells in S phase (35.2%) and G2/M phases (6.3%) of the cell cycle. When co-treated with panobinostat (PS), 200 nM to 1000nM DZNep is shown to decrease cell viability in OCI-AML3 and HL-60 cultures more effectively than DZNep alone, in a dose-dependent manner. DZNep (1mg/kg, twice per week, i.p.) and PS significantly prolong the survival of mice implanted with HL-60 cells compared to treatment with either compound alone. DZNep is also a known S-adenosyl-L-homocysteine (AdoHcy) hydrolase inhibitor.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
EZH2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 25 mM (1 mg/98 µL) sterile-filtered solution of Histone Methyltransferase EZH2 Inhibitor, DZNep (Cat. No. 252790) in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 monthst at -20°C.

Other Notes

Sun, F., et al. 2009. Mol. Cancer Ther.8, 3191.
Miranda, T.B., et al. 2009. Mol. Cancer Ther.8, 1579.
Fiscus, W., et al. 2009. Blood13, 2733.
Tan, J., et al. 2007. Genes Dev.21, 1050.
Chiang, P.K., et al. 1992. J. Biol. Chem.267, 4988.
Tseng, C.K., et al. 1989. J. Med. Chem.32, 1442.
Glazer, R.I., et al. 1986. Biochem. Biophys. Res. Commun.135, 688.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service