Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.06047

Millipore

3,3′-Diamino-4′-methoxyflavone, DD1

Synonym(s):

3,3′-Diamino-4′-methoxyflavone, DD1, 3-amino-2-(3-amino-4-methoxyphenyl)-4H-chromen-4-one, p76S6 Inhibitor, DD1, Proteasome Inhibitor, DD1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O3
Molecular Weight:
282.29
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

2 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable flavone derivative that arrests the proliferation of acute myeloid leukemia cells (NB4, U937, HL-60, OCI AML3), in a dose and time-dependent manner (optimum at ~20 µM and 96 h) and blocks the colony formation in samples from acute myeloid leukemia patients, without affecting normal blood cells. Shown to reduce mitochondrial membrane potential and Bad phosphorylation (at Ser136), induce Bax and caspase 3, 8, and 9 activation in U937 cells. Does not affect the phosphorylation of Akt1, but significantly reduces the expression and phosphorylation of p70S6 kinase in U937 cells. Reported to inhibit the chymotrypsin-like activity of proteasome (IC50 = 2.0 µM) and this effect is synergistic with bortezomib (Cat. No. 504314).

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Piedfer. M., et al, 2013. Biochim. Biophys. Acta.1833,1316
Quiney, C., et al, 2004. Leuk. Res.28, 851.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service