Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05854

Sigma-Aldrich

TGR5 Receptor Agonist, Cpd23g

Synonym(s):

TGR5 Receptor Agonist, Cpd23g, (4-cyclopropyl-3,4-dihydroquinoxalin-1(2H)-yl)(4-(2,5-dimethylphenoxy)pyridin-3-yl)methanone, Bile Acid Receptor GPBAR-1 Agonist, Bile Acid Receptor Agonist, G protein-coupled bile acid receptor 1 Agonist

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$306.00

$306.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$306.00

About This Item

Empirical Formula (Hill Notation):
C25H25N3O2
CAS Number:
Molecular Weight:
399.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$306.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

N2(CCN(c5c2cccc5)C(=O)c3cnccc3Oc4c(ccc(c4)C)C)C1CC1

InChI key

JQULIQJSYPZQMA-UHFFFAOYSA-N

General description

A cell-permeable, orally available, phenoxypyrimidine carboxamide derivative that acts as a highly potent and selective agonist of Protein coupled receptor TGR5 (EC50 = 720 pM and 6.2 nM for human and mouse TGR5, respectively). Does not exhibit any significant affinity towards other related targets such as GPR40, GPR119, and GPR120. Shown to increase glucagon-like peptide-1 (GLP-1) secretion in a dose-dependent manner and significantly reduce blood glucose levels in db/db mice following a single oral dose (50 mg/kg). Exhibits desirable pharmacokinetic properties in rodent models (t½ = 1.5 h, Cmax = 56 ng/ml; and AUC = 147 ng.h/ml following an oral dose of 5 mg/kg).

Biochem/physiol Actions

Primary Target
TGR5
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Duan, H., et al. 2012, J. Med. Chem.55, 10475.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service