Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05764

Sigma-Aldrich

NMDA Antagonist XI, N1-dansyl-spermine

Synonym(s):

NMDA Antagonist XI, N1-dansyl-spermine, NMDA channel blocker, N1-dansyl-spermine, N1-DnsSpm

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H37N5O2S · 3HCl
Molecular Weight:
545.01
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

potency

0.31 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)NCCCNCCCCNCCCN

General description

An N-sulfonyl-polyamine that acts as a potent and more specific antagonist of NMDA receptors expressed in Xenopus oocytes (IC50 = 310, 340 nM in NR1/NR2A and NR1/NR2B, respectively). The inhibition appears to be voltage-dependent and is non-competitive with respect to glutamate and glycine. Exhibits about 50-fold greater selectivity over NR1/NR2C and NR1/NR2D receptors (IC50 = 16 and 13 µM, respectively). Also shown to block c-AMP-dependent, calmodulin-activated phosphodiesterase (IC50 = 58 µM) and diminishes the induction of ornithine decarboxylase. At higher concentration (˜100 µM) it is shown to reduce spontaneous epileptiform activity in mouse cortical slices.

Biochem/physiol Actions

Primary Target
NMDA

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kirby, B.P., et al. 2005, Eur. J. Pharmacol. 524, 53.
Li, J., et al. 2005, Brain Res. 1055, 180.
Kirby, B. et al. 2004, Brain Res. 1011, 69.
Seiler, N., et al. 1998, J. Biochem. Cell Biol. 30, 393.

Chao, J., et al. 1997, Mol. Pharm. 51, 861.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service