5.05602
BDK Inhibitor II, (S)-CPP
Synonym(s):
BDK Inhibitor II, (S)-CPP, (S)-α-Chloro-phenylpropionic acid, BCKD Kinase Inhibitor II, Branched-Chain α-Ketoacid Dehydrogenase Kinase Inhibitor II, BCKD Kinase Inhibitor II, Branched-Chain α-Ketoacid Dehydrogenase Kinase Inhibitor II, (S)-α-Chloro-phenylpropionic acid
Select a Size
$197.00
Select a Size
About This Item
$197.00
Recommended Products
Assay
≥98% (HPLC)
Quality Level
form
oil
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated
protect from light
color
clear colorless
solubility
DMSO: 100 mg/mL
storage temp.
−20°C
SMILES string
Cl[C@@H](Cc1ccccc1)C(=O)O
InChI
1S/C9H9ClO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6H2,(H,11,12)/t8-/m0/s1
InChI key
LIDRHDRWTSPELB-QMMMGPOBSA-N
General description
Biochem/physiol Actions
BDK
Packaging
Warning
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service