Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05278

Sigma-Aldrich

PNU 282987

Synonym(s):

PNU 282987, PNU-282987, N-(3R)-1-Azabicyclo[2.2.2]oct-3-yl-4-chlorobenzamide hydrochloride, α7 nAChR Agonist, PNU 282987, alpha 7 Nicotinic Acetylcholine Receptor Agonist, PNU 282987, PNU282987

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17ClN2O · xHCl
CAS Number:
Molecular Weight:
264.75 (free base basis)
UNSPSC Code:
12352200

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

26 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

water: 100 mM

storage temp.

2-8°C

InChI

1S/C14H17ClN2O.ClH/c15-12-3-1-11(2-4-12)14(18)16-13-9-17-7-5-10(13)6-8-17;/h1-4,10,13H,5-9H2,(H,16,18);1H/t13-;/m0./s1

InChI key

HSEQUIRZHDYOIX-ZOWNYOTGSA-N

General description

A potent agonist highly selective for α7 nAChRs (Ki = 26 nM). Used in investigating the roles of α7 nAChRs in sensory gating, learning and memory, cognitive functions, and treatment of Schizophrenia.

Biochem/physiol Actions

Primary Target
α7 nAChRs
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hajós, M., et al., 2010, Curr. Pharm. Des.16, 538.
Bodnar, L., et al., 2005. J. Med. Chem.48, 905.
Hajós, M., et al., 2005. J. Pharmacol. Exp. Ther.312, 1213.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service