Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.05223

Sigma-Aldrich

CMP-Sialic Acid disodium salt

≥95% (HPLC), liquid, activated form of sialic acid, Calbiochem®

Synonym(s):

CMP-Sialic Acid, Disodium Salt InSolution, Cytidine-5ʹ-MonoPhospho-N-Acetylneuraminic acid, 2Na, CMP-N-Acetylneuraminic Acid, 2Na, CMP-NANA, CMP-Neu5Ac

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29N4O16P · 2Na
CAS Number:
Molecular Weight:
658.41
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

product name

CMP-Sialic Acid, Disodium Salt, InSolution, ≥95%, 25 mM aqueous solution, required for the biosynthesis of sialic acid-containing complex carbohydrates

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

storage temp.

−70°C

InChI

1S/C20H31N4O16P/c1-7(26)22-12-8(27)4-20(18(32)33,39-16(12)13(29)9(28)5-25)40-41(35,36)37-6-10-14(30)15(31)17(38-10)24-3-2-11(21)23-19(24)34/h2-3,8-10,12-17,25,27-31H,4-6H2,1H3,(H,22,26)(H,32,33)(H,35,36)(H2,21,23,34)

InChI key

TXCIAUNLDRJGJZ-UHFFFAOYSA-N

General description

CMP-sialic acid is the activated form of sialic acid that is required for the biosynthesis of sialic acid-containing complex carbohydrates. It is a widespread carbohydrate with numerous biological functions, including blood protein half-life regulation, variety of toxin neutralization, cellular adhesion and glycoprotein lytic protection.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 25 mM (10 mg/608 µL) sterile-filtered solution of CMP-Sialic Acid, Disodium Salt (Cat. No. 233264) in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 1 year at -70°C.

Other Notes

Schauer, R., et al. 1972. Hoppe Seylerʹs Z. Physiol. Chem.6, 353.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Enzymatic glycosyltransferase specificity challenges the one enzyme-one linkage concept.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service