Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05206

Sigma-Aldrich

Photochromic TrpA1 Channel Activator, Optovin

Synonym(s):

Photochromic TrpA1 Channel Activator, Optovin, (E)-5-((2,5-dimethyl-1-(pyridin-3-yl)-1H-pyrrol-3-yl)methylene)-2-thioxothiazolidin-4-one, PDSP 22594, Transient Receptor Potential A1 Ion Channel Activator I, TrpA1 Channel Activator I

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$204.00

$204.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$204.00

About This Item

Empirical Formula (Hill Notation):
C15H13N3OS2
CAS Number:
Molecular Weight:
315.41
MDL number:
UNSPSC Code:
41106609
NACRES:
NA.77

$204.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

form

powder

potency

2 μM EC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

S1C(=S)NC(=O)C1=Cc2c([n](c(c2)C)c3cnccc3)C

InChI

1S/C15H13N3OS2/c1-9-6-11(7-13-14(19)17-15(20)21-13)10(2)18(9)12-4-3-5-16-8-12/h3-8H,1-2H3,(H,17,19,20)

InChI key

YISGMOZSGOGYOU-UHFFFAOYSA-N

General description

A rhodanine derivative that acts as a reversible, photoactivated (λmax = 415 nm), and highly TrpA1-selective channel activator, presumably by a light-activated cysteine alkylation mechanism via the α,β-unsaturated alkene group of Optovin rhodanine moiety, and enables light-induced primary murine DRG calcium response in a TrpA1-dependent and HC-030031- (Cat. No. 648485) blockable manner, allowing light-induced activation only in hTrpA1-expressing, but not TrpV1- or TrpM8-expressing, HEK293T transfectants. Its in vivo efficacy is demonstrated in light-induced zebrafish motor activity (EC50/ECmax = 2 µM/3 µM; with >1.6 µW/min 387-405 nm excitation) and low power 405 nm laser-induced head shake/twitch response in mice (0.3 µmol Optovin/20 uL DMSO/ear applied atopically before laser illumination). Exhibits little or no inhibitory activity toward hERG or a panel of 50 human and rodent CNS receptors, channels, and transporters.

Biochem/physiol Actions

Primary Target
TrpA1

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Kokel, D., et al. 2013, Nat. Chem. Biol.9, 257.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service