Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05016

Sigma-Aldrich

CX546

Synonym(s):

CX546, AMPA Positive Allosteric Modulator, CX546, CX-546, CX 546

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO3
CAS Number:
Molecular Weight:
247.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (TLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: soluble

storage temp.

2-8°C

InChI

1S/C14H17NO3/c16-14(15-6-2-1-3-7-15)11-4-5-12-13(10-11)18-9-8-17-12/h4-5,10H,1-3,6-9H2

InChI key

LJUNPHMOGNFFOS-UHFFFAOYSA-N

General description

A positive allosteric modulator selective for the AMPA receptors. Used in studies related to Schizophrenia, neuroplasticity, Autism and neuronal differentiation.

Biochem/physiol Actions

Primary Target
AMPA

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Silverman, L., et al. 2013. Neuropharmacol.64, 268.
Peleg-Raibstein, D., et al. 2012. Curr. Antipsych.2012, 361.

Schitine, C., et al. 2012. Eur. J. Neurosci.35, 1672.
Lipina, T., et al. 2006. Neuropsychopharmacology.32, 745.
Arai, C., et al. 2004. Neurosci.123, 1011.
Nagarajan, N., et al. 2001. Neuropharmacol.41, 650.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service