Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.04904

Sigma-Aldrich

Ifenprodil Hemitartrate

Synonym(s):

Ifenprodil Hemitartrate, erythro-4-(2-(4-Benzylpiperidin-1-yl)-1-hydroxypropyl)phenol hemitartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27NO2 · 0.5C4H6O6
CAS Number:
Molecular Weight:
400.49
MDL number:
UNSPSC Code:
12352200

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

0.15 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mM

storage temp.

2-8°C

InChI

1S/2C21H27NO2.C4H6O6/c2*1-16(21(24)19-7-9-20(23)10-8-19)22-13-11-18(12-14-22)15-17-5-3-2-4-6-17;5-1(3(7)8)2(6)4(9)10/h2*2-10,16,18,21,23-24H,11-15H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1

InChI key

DMPRDSPPYMZQBT-CEAXSRTFSA-N

General description

A partial non-competitive antagonist highly selective for GluN1/N2B subtype NMDA receptors (IC50 = 0.15 µM, maximal inhibition ˜90%) over other subtypes (IC50 = 39, 29, 76 µM for GluN1/N2A, GluN1/N2C and GluN1/N2D subtype receptors, respectively). Frequently used in studies of various brain functions and disorders involving neural transmissions mediated by GluN1/N2B subtype NMDA receptors.
A partial non-competitive antagonist highly selective for GluN1/N2B subtype NMDA receptors (IC50 = 0.15 µM, maximal inhibition ˜90%) over other subtypes (IC50 = 39, 29, 76 µM for GluN1/N2A, GluN1/N2C and GluN1/N2D subtype receptors, respectively). Frequently used in studies of various brain functions and disorders involving neural transmissions mediated by GluN1/N2B subtype NMDA receptors.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
GluN1/N2B subtype NMDA receptors

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhang, H., et al. 2013. Mol. Brain.6, 13.
Kishimoto, A., et al. 2013. J. Clin. Psychopharmacol.33, 143.
Bhat, J., et al. 2013. Mol. Pharmacol.83, 9.
Amico-Ruvio, S., et al. 2012. 82, 1074, 81.
Bhattacharya, P., et al. 2012. Med. Hypotheses.79, 516.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service