Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.04840

Sigma-Aldrich

(R)-CPPene

Synonym(s):

(R)-CPPene, (R)-4-[(2E)-3-Phosphono-2-propenyl]-2-piperazinecarboxylic acid, SDZ EAA 494, Midafotel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15N2O5P
CAS Number:
Molecular Weight:
250.19
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

H2O: 100 mM

storage temp.

2-8°C

InChI

1S/C8H15N2O5P/c11-8(12)7-6-10(4-2-9-7)3-1-5-16(13,14)15/h1,5,7,9H,2-4,6H2,(H,11,12)(H2,13,14,15)/b5-1+/t7-/m1/s1

InChI key

VZXMZMJSGLFKQI-ABVWVHJUSA-N

General description

A highly potent competitive antagonist of NMDA receptors (ED50 = 39 nM; pA2 = 6.8). Biologically active admitted systemically. Frequently used in assessing the functional roles of NMDA receptors mediated neurotransmissions in a variety of research areas, such as excitotoxic cell death, apoptosis, bipolar disorders, ischemia, traumatic brain injury, epilepsy, and neuropathic pain.

Biochem/physiol Actions

Primary Target
NMDA receptors

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lowe, D., et al. 1990. Neurosci. Lett.113, 315.
Bullock, R., et al. 1990. Stroke.21, 32.
Bespalov, A., et al. 1998. Eur. J. Pharmacol.351, 299.

Lowe, D., et al. 1994. Neurochem. Inter.25, 583.
Duty, S. 2012. CNS Drugs26, 1017.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service