Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.04819

Sigma-Aldrich

Porcn Inhibitor III, IWP-L6

Synonym(s):

Porcn Inhibitor III, IWP-L6, IWPL6, Wnt Pathway Inhibitor XIX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20N4O2S2
CAS Number:
Molecular Weight:
472.58
UNSPSC Code:
12352200

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

InChI

1S/C25H20N4O2S2/c30-22(28-21-12-11-18(15-26-21)17-7-3-1-4-8-17)16-33-25-27-20-13-14-32-23(20)24(31)29(25)19-9-5-2-6-10-19/h1-12,15H,13-14,16H2,(H,26,28,30)

InChI key

QESQGTFWEQMCMH-UHFFFAOYSA-N

General description

A cell-permeable pyridinyl-phenylacetamide that is reported to be 60-times more potent than IWP-2 (Cat. No. 681671) in suppressing autonomous Wnt signaling in murine L-Wnt-STF fibroblasts (IC50 = 0.5 nM) and 100-times more potent in blocking branching morphogenesis in cultured murine embryonic kidney (complete blockage at 50 nM). Also shown to be at least twice as potent as TNKS inhibitor IWR-1 (Cat. No. 681669) in preventing the regeneration of severed zebrafish tailfin and in inhibiting zebrafish embryo prosterior axis formation (Effective conc = 5 µM). Its susceptibility to metabolization by serum carboxylesterase limits its in vivo use in rats and mice.
A cell-permeable, benzothiazole-to-5-phenylpyridin substituted IWP-2 (Cat. No. 681671) derivative that is reported to be 60-times more potent than IWP-2 in suppressing autonomous Wnt signaling in murine L-Wnt-STF fibroblasts (IC50 = 0.5 vs. 30 nM) and 100-times more potent in blocking Wnt signaling-mediated branching morphogenesis in cultured murine embryonic kidneys (complete blockage with 50 nM IWP-L6 or 5 µM IWP-2). Also shown to be at least twice as potent as TNKS inhibitor IWR-1 (Cat. No. 681669) in preventing the regeneration of severed zebrafish tailfin and in inhibiting zebrafish embryo prosterior axis formation (Effective conc = 5 µM). Its susceptibility to metabolization by serum carboxylesterase limits its in vivo use in rats and mice (t1/2 = 2 and 190 min, respectively, in murine or rat plasma).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Porcupine
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wang, X., et al. 2013. J. Med. Chem.56, 2700.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service