Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.04596

Sigma-Aldrich

TCN-201

Synonym(s):

TCN-201, 3-Chloro-4-fluoro-N-[(4-{[2-(phenylcarbonyl)hydrazino]carbonyl}phenyl)methyl]benzenesulfonamide, NMDA Antagonist X

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17ClFN3O4S
CAS Number:
Molecular Weight:
461.89
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

InChI

1S/C21H17ClFN3O4S/c22-18-12-17(10-11-19(18)23)31(29,30)24-13-14-6-8-16(9-7-14)21(28)26-25-20(27)15-4-2-1-3-5-15/h1-12,24H,13H2,(H,25,27)(H,26,28)

InChI key

FYIBXBFDXNPBSF-UHFFFAOYSA-N

General description

A negative allosteric modulator selective for NR2A containing NMDA receptors (pIC50 = 6.8 and <4.3 µM for NR2A and NR2B, respectively). No effect on NR2B-containing NMDA receptors with concentrations less than 50 mM. Inhibition on NR2A containing NMDA receptors is glycine (or D-serine)-binding, but not glutamate-binding, dependent.

Biochem/physiol Actions

Primary Target
NMDA receptors

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bettini. E., et al. 2010. J. Pharm. Exp. Ther.335, 644.
Shin, A., et al. 2011. Br. J. Pharmacol.166, 1002.
Edman, S., et al. 2012. Neuropharmacology63, 441.
Hansen, K., et al. 2012. J. Neurosci.32, 6197.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service