Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.04570

Sigma-Aldrich

Tunicamycin, Streptomyces lysosuperficus

InSolution, 25 mM in DMSO

Synonym(s):

InSolution Tunicamycin, Streptomyces lysosuperficus, NSC 177382

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H64N4O16
CAS Number:
Molecular Weight:
844.94
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

InChI

1S/C30H46N4O16/c1-11(2)5-4-6-16(38)32-19-23(43)20(40)14(47-29(19)50-28-18(31-12(3)36)22(42)21(41)15(10-35)48-28)9-13(37)26-24(44)25(45)27(49-26)34-8-7-17(39)33-30(34)46/h4,6-8,11,13-15,18-29,35,37,40-45H,5,9-10H2,1-3H3,(H,31,36)(H,32,38)(H,33,39,46)/b6-4+/t13?,14-,15-,18-,19-,20+,21-,22-,23-,24+,25-,26-,27-,28-,29-/m1/s1

InChI key

ZHSGGJXRNHWHRS-PEALBESXSA-N

Biochem/physiol Actions

Cell permeable: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 25 mM (5 mg/237 µL) sterile-filtered solution of Tunicamycin, Streptomyces lysosuperficus (Cat. No. 654380).in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -7-°C.



Aqueous solutions can be prepared from stock solutions in DMSO by diluting with H₂O at pH 8.0 to 10.0 or with appropriate buffers at pH >7.0 (preferably >8.0).

Other Notes

Tordai, A., et al. 1995. Biochem. Biophys. Res. Commun.206, 857.
Price, B.D., et al. 1992. J. Cell Physiol.152, 545.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service