Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.04041

Sigma-Aldrich

CaCC Blocker IV, Benzbromarone

Synonym(s):

CaCC Blocker IV, Benzbromarone, CaCC Blocker IV, Calcium-Activated Chloride Channel Blocker IV, TMEM16 Blocker IV, (3,5-dibromo-4-hydroxyphenyl)(2-ethylbenzofuran-3-yl)methanone, ANO Blocker IV, Anoctamin Blocker IV, BBR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12Br2O3
CAS Number:
Molecular Weight:
424.08
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A cell-permeable uricosuric agent that, in addition to its known efficacy in gout treatment, acts as a reversible CaCC blocker and is reported to be more potent than NFA (Cat. No. 481987) and NPPB (Cat. No. 484100) in suppressing 500 nM Ionomycin- (Cat. Nos. 407950, 407952, and 407953) induced I- influx in TMEM16A-expressing HEK293 cells (IC50 = 9.97, 140, and 150 µM, respectively) as well as in blocking voltage-gated Cl- current in TMEM16B-expressing HEK293 (64% inhibition with 10 µM BBR vs. 47% inhibition with 300 µM NFA; by whole cell patch clamp), while exhibiting much reduced potency against voltage-gated current via ENaC (α, β, and γ) or CFTR (by 25% and 17% with 10 µM BBR, respectively, using HEK293 ENaC or CFTR transfectants). Shown to significantly reduce 100 µM ATP-induced CaCC current (IAC current = 1.137 vs. 3.099 µA/cm2 with or without 10 µM BBR) and mucin secretion from NHBE (normal human bronchial epithelial) as well as suppress human ASM (airway smooth muscle) contraction upon muscarinic receptor stimulation.
A cell-permeable uricosuric agent that, in addition to its known efficacy in gout treatment, acts as a reversible CaCC blocker and is reported to be more potent than NFA (Cat. No. 481987) and NPPB (Cat. No. 484100) in suppressing 500 nM Ionomycin- (Cat. Nos. 407950, 407952, and 407953) induced I- influx in TMEM16A-expressing HEK293 cells (IC50 = 9.97, 140, and 150 µM, respectively) as well as in blocking voltage-gated Cl- current in TMEM16B-expressing HEK293 cells, while exhibiting much reduced potency against ENaC (α, β, and γ) or CFTR. Shown to significantly reduce mucin secretion from NHBE (normal human bronchial epithelial) and suppress human ASM (airway smooth muscle) contraction upon muscarinic receptor stimulation.

Biochem/physiol Actions

Primary Target
TMEM16A-CaCC
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Kobayashi, K., et al. 2012. Drug Metab. Pharmacokinet. In press.
Zhang, C.H., et al. 2013. Am. J. Respir. Crit. Care Med.187, 374.
Huang, F., et al. 2012. Proc. Natl. Acad. Sci. USA.109, 16354.
Fujimori, S., et al. 2011. Nucleosides Nucleotides Nucleic Acids30, 1035.
Perez-Ruiz, F., et al. 1998. Ann. Rheum. Dis.57, 545.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service