Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.00523

Sigma-Aldrich

ASIC Channel Modulator II, GMQ

Synonym(s):

ASIC Channel Modulator II, GMQ, 2-guanidine-4-methylquinazoline, Acid-Sensing Ion Channel Modulator II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N5
CAS Number:
Molecular Weight:
201.23
UNSPSC Code:
12352200

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 16.6 mg/mL

storage temp.

2-8°C

General description

A positively charged guanidium-containing quinazoline that modulates ASIC function by blocking acid-induced maximal peak current as well as by altering ASICs pH dependence for activation and inactivation, enabling a partial, but sustained ASIC3 activation around neutral pH (4.4% of pH 5-induced peak current with 1 mM GMQ at pH 7.4), which is inhibitable by Amiloride (Cat. No. 129876). Intraplantar paw injection (10 µL 100 µM) is reported to elicit a pain response in asic3+/+, but not asic3-/-, mice in vivo. GMQ targeting area appears to overlap with Amiloride binding site in the pore region, but is distinct from ASIC calcium binding site.

Biochem/physiol Actions

Primary Target
ASIC3
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Alijevic, O., and Kellenberger, S. 2012. J Biol Chem.287, 36059.

Sherwood, T.W., et al. 2012. Am. J. Physiol. Cell Physiol.303, C699.
Yu, Y., et al. 2010. Neuron.68, 61.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service