Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.00505

Sigma-Aldrich

CRM1 Inhibitor IV, KPT-251

Synonym(s):

CRM1 Inhibitor IV, KPT-251, KPT251, (Z)-2-(2-(3-(3,5-bis(trifluoromethyl)phenyl)-1H-1,2,4-triazol-1-yl)vinyl)-1,3,4-oxadiazole, Exportin 1 Inhibitor IV, Chromosome Region Maintenance 1 Protein Inhibitor IV, CRM1 Inhibitor IV, XPO1 Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H7F6N5O
CAS Number:
Molecular Weight:
375.23
UNSPSC Code:
12352200

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 25 mg/mL

storage temp.

−20°C

General description

A cell-permeable, irreversible, triazole-containing heterotricyclic CRM1 inhibitor that effectively blocks CRM1-mediated nuclear export (4 h 1µM in U2OS cells) via covalent interaction with CRM1 NES- (nuclear export signal) binding groove cysteine (Cys528 in human). Exhibits antileukemic activity against primary human CD19+ CLL (IC50<500 nM) via apoptosis induction even in the presence of stroma cells or other known CLL survival stimuli, while exhibiting little cytotoxicity toward CD56+ NK cells from healthy donors (1 µM; up to 72 h). Reported to prolong the survival of SCID mice (75 mg/kg p.o.) engrafted with murine Eμ-TCL1 or human MV4-11 leukemia in vivo.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
A cell-permeable, irreversible, triazole-containing heterotricyclic CRM1 inhibitor that effectively blocks CRM1-mediated nuclear export (complete HIV-1 Rev nuclear retention after 4 h 1µM incubation in RevGFP-expressing U2OS cells) via covalent interaction with Cys528 (numbering based on human sequence) in the CRM1 NES- (nuclear export signal) binding groove, while being ineffective against CRM1C528S-Rev interaction in RevBFP-expressing HeLa cells. Exhibits antileukemic activity in primary human CD19+ CLL (chronic lymphocytic leukemia) cultures (IC50<500 nM) via apoptosis induction even in the presence of stroma cells or other known CLL survival stimuli, while exhibiting little cytotoxicity toward CD56+ NK cells from healthy donors (1 µM; up to 72 h). KPT-251 is orally available in mice (tmax = 0.5 h; t1/2 = 3.83 h; Cmax = 439 ng/mL; AUCINF = 1590 h·ng/mL; 10 mg/kg p.o.) and reported to prolong the survival of SCID mice (75 mg/kg p.o.) engrafted with murine Eμ-TCL1 (mean survival time = 130.5 d vs. 72 d, respectively, with or without treatment) and human MV4-11 leukemia (62% survival rate on d 60 vs. 24% on d 34, respectively, with or without treatment) in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
XPO1
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Etchin, J., et al. 2013. Leukemia.27, 66.
Lapalombella, R., et al. 2012. Blood.120, 4621.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service