Skip to Content
MilliporeSigma

5.00299

SAFC

Nicotinamide

EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, USP, FCC

Pharma Manufacturing

Synonym(s):

Nicotinamide, Nicotinamide, Niacinamide, 3-Pyridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
MDL number:
UNSPSC Code:
12352111
EC Index Number:
202-713-4

Agency

BP
JP
Ph. Eur.
USP

Quality Level

reg. compliance

FCC

description

EMPROVE® ESSENTIAL Ph Eur,BP,USP,JP,FCC

product line

EMPROVE® ESSENTIAL

form

solid

autoignition temp.

480 °C

potency

3530-3540 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

technique(s)

API processing | cocrystal formation: suitable

pH

6.0-7.5 (20 °C, 50 g/L in H2O)

bp

150-160 °C/0.0007 hPa

mp

128-131 °C (sublimed)

transition temp

flash point 182 °C

density

1.40 g/cm3 at 20 °C

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

InChI

1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)

InChI key

DFPAKSUCGFBDDF-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Nicotinamide is a commonly used ingredient in food and pharmaceutical applications. In pharmaceutical formulations, nicotinamide has been used for a range of different dosage forms.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service